Titer-Tops, Electron Microscopy Sciences | Avantor
Titer-Tops, Electron Microscopy Sciences
Titer-Tops, Electron Microscopy Sciences
Certificates
Titer-Tops, Electron Microscopy Sciences
Titer-Tops, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Titer-Tops prevents sample evaporation, well to well contamination, external contamination and sample spillage. Titer-Tops are made from 4.4 mil polyethylene film with an FDA approved non cytotoxic acrylic adhesive.
  • Titer-Tops are chemically resistant and thermally stable
  • Titer-Tops are gamma sterile
  • They are easily pierced with a micropipette tip to provide access to sample wells
Each pre-cut rectangular sheet is 6 x 3.25" with a paper liner. The center section of the liner measures 5 x 3.25" and once it is peeled off it leaves two hand-grip extension tabs on the two ends. These tabs allow for easy application of the Titer-Top to the plate. The acrylic adhesive is waterproof, solvent resistant, and compatible with all plastic microtiter plates. Titer-Tops will stay in place even when exposed to 100% humidity and temperatures ranging from -40 to 100 °C.
Product Family Options
Informations sur le produit
  • Désignation
    Épaisseur
    L×l
    Availability
    Price
  • Titer-Tops, 50
    112 µm
    6×3.25"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $102.91
    $102.91
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $93.36
    $93.36
    Each
       $102.91
    Pack of 50
       $93.36
    Supplier Number: 70538-05
  • Titer-Tops, 100
    112 µm
    6×3.25"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $232.93
    $232.93
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $177.23
    $177.23
    Each
       $232.93
    Pack of 100
       $177.23
    Supplier Number: 70538-10