Tissue Processing Capsules, Electron Microscopy Sciences
Tissue Processing Capsules, Electron Microscopy Sciences
Tissue Processing Capsules, Electron Microscopy Sciences
Certificates
Tissue Processing Capsules, Electron Microscopy Sciences
Tissue Processing Capsules, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These one piece plastic capsules are hinged and perfectly aligned, offering convenience and ease of use
Both the top and the bottom of the capsule have an open mesh offering maximum fluid flow during processing. A frosted marking surface assures permanent identification. These capsules are commonly used with the Embedding Ring System. They are available in two sizes and five colors.
Product Family Options
Product Information
  • Désignation
    Color
    Ø
    Profondeur
    Availability
    Price
  • Tissue Processing Capsule for Normal Tissue
    White
    38 mm
    8 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $358.78
    $358.78
    Log in for availability
    Pack of 500
    Pack of 500 - $283.29
    $283.29
    Each
       $358.78
    Pack of 500
       $283.29
    Supplier Number: 62358-W
  • Tissue Processing Capsule for Biopsy Tissue
    White
    29 mm
    6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $361.88
    $361.88
    Log in for availability
    Pack of 500
    Pack of 500 - $328.31
    $328.31
    Each
       $361.88
    Pack of 500
       $328.31
    Supplier Number: 62359-W