Tissue Mounting Stubs, Electron Microscopy Sciences
Tissue Mounting Stubs, Electron Microscopy Sciences
Aluminum Tissue Mounting Stub
Catalog # 100490-730
Supplier #:  70145-05
Tissue Mounting Stubs, Electron Microscopy Sciences
Catalog # 100490-730
Tissue Mounting Stubs, Electron Microscopy Sciences
Tissue Mounting Stubs, Electron Microscopy Sciences
Aluminum Tissue Mounting Stub
Price
$259.77
Log in for availability
Each Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Machined from aluminum rods with one end tapered to 4.8 mm in diameter. The specimen can be oriented and mounted to the tip with glue and it is ready for sectioning. The body size is "00" and it measures 11 mm long.
Specifications
  • Désignation:
    Aluminum Tissue Mounting Stub