Thin Sample Holder, Electron Microscopy Sciences
Thin Sample Holder, Electron Microscopy Sciences
Thin Sample Holder, Electron Microscopy Sciences
Certificates
Thin Sample Holder, Electron Microscopy Sciences
Thin Sample Holder, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals and more.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Thin sample holder, 8 mm Pin
    Log in for availability
    Log in for availability
    Each
    Each - $130.41
    $130.41
    Each
       $130.41
    Supplier Number: 75948-08
  • Thin sample holder, 15 mm pin
    Log in for availability
    Log in for availability
    Each
    Each - $146.83
    $146.83
    Each
       $146.83
    Supplier Number: 75948-15