ContactUsLinkComponent
MAC Thin Foils Standards for STEM, Electron Microscopy Sciences
Catalog # 100495-262
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
MAC Thin Foils Standards for STEM, Electron Microscopy Sciences
Catalog # 100495-262
Supplier Number:  80082-25
Minority or Woman-Owned Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pack type:
    25/set
  • Description:
    Thin Foil Standards for STEM
  • Cat. no.:
    100495-262
  • Supplier No.:
    80082-25
Specifications
About this item
Selection of 25 high purity thin foils, 0.1 mm thick, 3 mm Ø to fit into TEM grid holders for use in the STEM mode.
  • Polished to a 3 μm diamond finish
Come in a variety of pure metals, including Aluminum, Cadmium, Cobalt, Copper, Gold, Hafnium, Indium, Iron, Magnesium, Molybdenum, Nickel, Niobium, Palladium, Platinum, Rhodium, Silver, Tantalum, Tin, Titanium, Tungsten, Vanadium, Yttrium, Zinc, Zirconium and AISI 316 S/S.