The Original LKB Grid Storage Box, Electron Microscopy Sciences
The Original LKB Grid Storage Box, Electron Microscopy Sciences
The Original LKB Grid Storage Box, Electron Microscopy Sciences
Certificates
The Original LKB Grid Storage Box, Electron Microscopy Sciences
The Original LKB Grid Storage Box, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The box  consists of 100 diamond shaped holes for storing up to 100 EM grids, either 3.05 or 2.3 mm in diameter.
  • The box measures L x W x H 75 x 55 x 7 mm (3 x 2 1/8 x 1/4")
  • It comes complete with 10 index cards
For years, The LKB Grid Storage Box is the one that everyone is looking for. Now it is available again from EMS. The box is made from ABS (a copolymer of Acrylonitrile, Butadien and Styrene) which will not tolerate temperatures above 70 °C, while the lid is made of  Polymethacrylate (Flexiglas, Perspex), which should not be exposed to temperatures above 45 °C. Neither the box or the lid will resist organic solvents.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • LKB 100 Grid Box
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,060.43
    $1,060.43
    Log in for availability
    Pack of 10
    Pack of 10 - $878.39
    $878.39
    Each
       $1,060.43
    Pack of 10
       $878.39
    Supplier Number: 71147-12
  • LKB 100 Grid Box
    Log in for availability
    Log in for availability
    Each
    Each - $98.67
    $98.67
    Each
       $98.67
    Supplier Number: 71147-01