ContactUsLinkComponent
EMS Staining Plate for Electron Microscopy, Electron Microscopy Sciences
  100491-794
Minority or Woman-Owned Business Enterprise
undefined
EMS Staining Plate for Electron Microscopy, Electron Microscopy Sciences
  100491-794
 :  71568
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Staining Plate for Electron Microscopy
  • Cat. no.:
    100491-794
  • Supplier No.:
    71568
The EMS Staining Plate for electron microscopy post-embedding staining and immunohistochemistry offers several advantages over all other commercially available staining devices
The EMS Staining plate for Electron Microscopy was developed by Dr. Miguel Berrios, at SUNY at Stony Brook, Dept of Pharmacological Sciences, School of Medicine, New York. The chemical etching process, antibody incubations and final staining with heavy metal salts of each grid is performed in the small cone-shaped wells on the EMS staining plate.
The EMS Staining Plate for electron microscopy post-embedding staining and immunohistochemistry offers several advantages over all other commercially available staining devices. The base plate is a solid piece of chemical-resistant silicone 127.5 mm long, 85.5 mm wide, 11.5 mm thick. With 96 cone-shaped wells organized (like the microtitration plate) in parallel rows of eight, the plate uses the lid of a 96-well Falcon 3072 Microtest™ III Plate as a cover. The base has two notches to serve for orientation and a 1.5 x 4.4 mm deep lip where the cover rests. Each well is an inverted cone 7 mm in diameter and 2 mm deep. Grids either float or rest at the bottom of each well. The wells allow incubation of a grid in 12-60 microliters of solution without reagent loss due to adsorption or cross contamination, even when the plate is tilted up at 70°. Due to the shape of the well, the flat surface of the grids never come into contact with the walls of the well, both facilitating sample staining and grid recovery. Plates made from silicone offer two advantages. Resistant to all chemicals and solvents. During manipulation of the grids in the well there is no risk of damaging the fine points of the tweezers.
Reference: Berrios, Miguel; (1991), A Staining Plate For Electron Microscopy. 48: 90-92.
Reference: Berrios, Miguel; (1991), A Staining Plate For Electron Microscopy. 48: 90-92.
The base plate is a solid piece of chemical-resistant silicone 127.5 mm long, 85.5 mm wide, 11.5 mm thick. With 96 cone-shaped wells organized (like the microtitration plate) in parallel rows of eight, the plate uses the lid of a 96-well Falcon 3072 Microtest™ III Plate as a cover. The base has two notches to serve for orientation and a 1.5 x 4.4 mm deep lip where the cover rests. Each well is an inverted cone 7 mm in diameter and 2 mm deep. Grids either float or rest at the bottom of each well. The wells allow incubation of a grid in 12-60 microliters of solution without reagent loss due to adsorption or cross contamination, even when the plate is tilted up at 70°. Due to the shape of the well, the flat surface of the grids never come into contact with the walls of the well, both facilitating sample staining and grid recovery. Plates made from silicone offer two advantages. Resistant to all chemicals and solvents. During manipulation of the grids in the well there is no risk of damaging the fine points of the tweezers.