The Embedding Capsule Substitute, Electron Microscopy Sciences
The strainers are available in the sizes of 35, 40, 70 and 100 microns.
- It is easier to see inside the well when centering the specimen in the tip
- It is easier to remove the block from the well
- There is no need for razor blades or press devices
The finished blocks have finer tips so when you trim deeper into the block the face doesn't increase in size rapidly; therefore, constant retrimming is not necessary.
- Désignation:Embedding Capsule Substitute