PTFE Beakers, Electron Microscopy Sciences
À propos de cet article
These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.
- With pouring spout
- Virtually immune to chemical attack
Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.
19 Options Available Below
Product Details & Documents
These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.
- With pouring spout
- Virtually immune to chemical attack
Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.
Product Family Options
Informations sur le produit
- CapacitéDésignationColorHauteurØ ext.AvailabilityPrice
- Supplier Number: 60936
- Supplier Number: 60938
- Supplier Number: 60940
- Supplier Number: 60941
- Supplier Number: 60942
- Supplier Number: 60947
- Supplier Number: 60946
- 500 mlPTFE BeakersWhite125 mm73 mmDiscontinued-Supplier Number: 60948
- 600 mlPTFE BeakerWhite127 mm80 mmDiscontinued-Supplier Number: 60949-06
- Supplier Number: 60949-10
- Supplier Number: 60949-20
- Supplier Number: 60936-C
- Supplier Number: 60938-C
- Supplier Number: 60940-C
- Supplier Number: 60942-C
- Supplier Number: 60944-C
- Supplier Number: 60946-C
- Supplier Number: 60947-C
- Supplier Number: 60948-C
- Supplier Number: 60949-06C
- 1000 mlPTFE Beaker CoverWhiteDiscontinued-Supplier Number: 60949-10C
- Supplier Number: 60941-C