Syringe Filtration System, Electron Microscopy Sciences
Syringe Filtration System, Electron Microscopy Sciences
Certificates
Syringe Filtration System, Electron Microscopy Sciences
Syringe Filtration System, Electron Microscopy Sciences
10 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Syrings filters with high burst strength for assured performance. Manufactured to ISO 9002 and GMP standard and 100% integrity tested.
  • For tissue culture and HPLC applications
  • Membrane choices include:
  • Regenerated cellulose (RC)
  • Polyethersulfone (PES)
  • Surfactant-free cellulose acetate (SFCA)
  • Nylon (NY)
  • 'PTFE' fluourocarbon resin
Syringe filter sizes include 4, 15, 25, or 26 mm diameter to handle volumes ranging from 0.5 to 100 ml. The 50 mm diameter 'PTFE' syringe filter are ideal for protecting the vacuum line and pump. These disposable filter units are ideal for tissue culture media, buffer solution, stain solution, biological fluids and most other aqueous solutions and reagents that are used daily in the EM lab.
Pore size and membrane type are printed on every syringe and the filters are certified non-cytotoxic and non-pyrogenic.
 : Sterile syringe filters are individually blister-wrapped in cases of 50. Non sterile syringe filters are packaged in cases of 50. The 50 mm 'PTFE' syringe filter is supplied in a case of 12.
Product Family Options
Filter Units with Polypropylene Housing
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Matériau du boîtier
    Availability
    Price
  • Syringe Filters RC
    4 mm
    0.20 µm
    PP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $431.90
    $431.90
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $333.14
    $333.14
    Each
       $431.90
    Pack of 50
       $333.14
    Supplier Number: 67000
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100489-792
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter RC
    15 mm
    0.20 µm
    PP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $325.38
    $325.38
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $349.16
    $349.16
    Each
       $325.38
    Pack of 50
       $349.16
    Supplier Number: 67002
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100502-764
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter RC
    25 mm
    0.20 µm
    PP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $563.04
    $563.04
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $439.08
    $439.08
    Each
       $563.04
    Pack of 50
       $439.08
    Supplier Number: 67002-25
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-292
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter NY
    25 mm
    0.20 µm
    PP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $607.72
    $607.72
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $482.34
    $482.34
    Each
       $607.72
    Pack of 50
       $482.34
    Supplier Number: 67018
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-254
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter NY
    25 mm
    0.45 µm
    PP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $599.36
    $599.36
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $476.12
    $476.12
    Each
       $599.36
    Pack of 50
       $476.12
    Supplier Number: 67019
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-256
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter "PTFE"
    50 mm
    0.20 µm
    PP
    Discontinued
    Supplier Number: 67022-50
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100490-670
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter "PTFE"
    25 mm
    0.45 µm
    PP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $596.38
    $596.38
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $431.23
    $431.23
    Each
       $596.38
    Pack of 50
       $431.23
    Supplier Number: 67025-25
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-294
    Minority or Woman-Owned Business Enterprise
Filter with Acrylic Housing
Informations sur le produit
  • Désignation
    Ø
    Taille de pore
    Matériau du boîtier
    Availability
    Price
  • Syringe Filter PES
    26 mm
    0.20 µm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $479.35
    $479.35
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $513.72
    $513.72
    Each
       $479.35
    Pack of 50
       $513.72
    Supplier Number: 67004
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100489-794
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter SFCA
    26 mm
    0.20 µm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $606.35
    $606.35
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $481.27
    $481.27
    Each
       $606.35
    Pack of 50
       $481.27
    Supplier Number: 67005
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-246
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter SFCA
    26 mm
    0.45 µm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $656.42
    $656.42
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $520.70
    $520.70
    Each
       $656.42
    Pack of 50
       $520.70
    Supplier Number: 67006
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-248
    Minority or Woman-Owned Business Enterprise
  • Syringe Filter SFCA-PF
    26 mm
    0.20 µm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $454.50
    $454.50
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $487.05
    $487.05
    Each
       $454.50
    Pack of 50
       $487.05
    Supplier Number: 67008
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100501-252
    Minority or Woman-Owned Business Enterprise