Stop Watch: Large Display, Electron Microscopy Sciences
Product Details & Documents
Very accurate with a click stop control button.
- Large LCD display
- Rugged ABS plastic case
It counts up to 60 minutes in ¹/₁₀₀th second resolution. It also has a split interval clock and calculator functions. Measures: 3×2½×¼".
Weight: 2.6 oz.
Delivery information: Comes complete with a leatherette case.
Spécifications
- Désignation:Stopwatch