Steinheil Magnifier, Electron Microscopy Sciences | Avantor
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, Electron Microscopy Sciences
Certificates
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The magnifier helps the user eliminate bulky and heavier conventional magnifiers.
A biconvex lens. Pocket size. The sliding frame is designed for the storage of the lens after use. It is good for the preliminary inspection of microscopic specimens. It is lightweight and has excellent chromatic correction with a reasonably wide field of view. Size: 39 x 24 x 22 mm. Average weight: 20 g.
Product Family Options
Informations sur le produit
  • Désignation
    Grossissement
    Availability
    Price
  • Steinheil Magnifier
    14×
    Discontinued
    Supplier Number: 68147
  • Steinheil Magnifier
    10×
    Discontinued
    Supplier Number: 68146
  • Steinheil Magnifier
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $152.36
    $152.36
    Each
       $152.36
    Supplier Number: 68145
  • Steinheil Magnifier
    20×
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $258.61
    $258.61
    Each
       $258.61
    Supplier Number: 68148