Steel Ruler, Electron Microscopy Sciences | Avantor
Steel Ruler, Electron Microscopy Sciences
Ruler, Steel
Catalog # 100502-102
Fournisseur #:  72909
Steel Ruler, Electron Microscopy Sciences
Catalog # 100502-102
Steel Ruler, Electron Microscopy Sciences
Steel Ruler, Electron Microscopy Sciences
Ruler, Steel
Price
$35.77
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel ruler.
  • Measures 1 mm thick, 1 cm wide, 1 dm long
  • Its volume is 1 cubic centimeter and its weight in grams is the specific gravity
  • One edge is graduated in millimeters
Spécifications
  • Désignation:
    Ruler
  • Matière:
    Steel