Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Certificates
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This hook and loop wrist strap is designed for those whom need to be static free in the working area.
This hook and loop wrist trap has a quick and easy adjustment. With a 1" wide conductive material that is sewn in and has a 1 MΩ resistor.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • 6' Coil Ground Cord Only,¼" Molded Female Snap
    Log in for availability
    Log in for availability
    Each
    Each - 40,97 $
    40,97 $
    Each
       40,97 $
    Supplier Number: 60622-30
  • Static Control Adjustable Wrist Band, 12' Coil Cord
    Discontinued
    Supplier Number: 60622-12