About this item
Cryogenics Liquid Nitrogen Transfer Vessels. Thermo-Flask provides safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2, and thus have a broad range of applications in industrial, educational, and clinical fields. Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1 x 10-5 mm of Hg to provide maximum coolant retention.
- Convenient dish-shaped design is ideal for bench top use with crushed ice, dry- ice, or alcohol.
- Also great as a hot liquid container.
- Stainless steel jacket and cushioned base (lids not included) will provide years of trouble-free use.
- Excellent for use as a cold trap.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 100497-524
- 100501-624
- Capacity:
- 1 L
- 1.9 L