Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences | Avantor
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
Certificates
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
Stainless Steel Forceps - Serrated Tips, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The forceps are made from stainless steel and have medium points, serrated tips and alignment pin.
  • With curved or straight tips
  • Available in 4" and 6" long
  • Curved type A tips comes with locking device
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Configuration
    Availability
    Price
  • Forceps, 2×1 mm Slot
    Stainless steel
    4"
    Straight Tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.05
    $45.05
    Each
       $45.05
    Supplier Number: 72991-4S
  • Forceps, 2×1 mm Slot
    Stainless steel
    4"
    Curved Tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $52.42
    $52.42
    Each
       $52.42
    Supplier Number: 72991-4C
  • Forceps, 2×1 mm Slot
    Stainless steel
    6"
    Curved "A" Tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.05
    $45.05
    Each
       $45.05
    Supplier Number: 72992-6C