Staining Rack; Coverglass, Electron Microscopy Sciences | Avantor
Staining Rack; Coverglass, Electron Microscopy Sciences
Large Coverglass Staining Rack, 10.2 cm (4")
Catalog # 100502-856
Fournisseur #:  72239-04
Staining Rack; Coverglass, Electron Microscopy Sciences
Catalog # 100502-856
Staining Rack; Coverglass, Electron Microscopy Sciences
Staining Rack; Coverglass, Electron Microscopy Sciences
Large Coverglass Staining Rack, 10.2 cm (4")
Price
$586.74
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from polished stainless steel.
  • Securely holds 30, 22 or 24 mm cover glass
  • Welded stainless steel
  • Spring-type detachable handle
  • 10.2 cm (4") long
Spécifications
  • Désignation:
    Large Coverglass Staining Rack
  • Longueur:
    10.2 cm (4")