Specimen Holders For SEM, Electron Microscopy Sciences
Specimen Holders For SEM, Electron Microscopy Sciences
Specimen Holders For SEM, Electron Microscopy Sciences
Certificates
Specimen Holders For SEM, Electron Microscopy Sciences
Specimen Holders For SEM, Electron Microscopy Sciences
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specimen holders are designed to improve your productivity and allow to view more than one sample at a time. This will save pump down time, keep the chamber cleaner and get more work done.
All mounts are machined from solid aluminum and come with spring clips/or setscrews to hold your specimens securely and are made to fit onto your stage and are designed to fit through all standard specimen exchange ports, and have a center-threaded port to accept the Adapter Pins
Universal SEM Sample Holder will hold almost any sample from 3 to 29 mm in diameter plus various odd shaped samples, which one of their dimensions is not greater than 29 mm. The samples are easily inserted or removed from the holder. The holder is made from aluminum and is supplied with four removable sample arms so that it can hold very small samples as well, and it provides good electrical contact to the stage. The AMRAY base is the standard base and it measures: 48 x 42 x 12 mm Thick. For all other makes and models, adapter Pin can choose to screw on to the AMRAY base holder.
Vertical Mount for thin samples flat base is designed to hold thin samples vertically in the SEM or any microscope. It is 25 mm in diameter and 10 mm thick. Each of the two loader jaws can hold up to 3 mm thick samples. The spring loader keeps thin samples vertical so that cross sections can be studied. This holder is very useful for cross sections of silicon wafers or multilayer capicators. This holder can be adapted to any adapter pin listed above to accommodate your instrument.
The Multi Pin Holder is designed to save time. It accommodates 3 or 5 of 12.5 mm (1/2") diameter surface, 1/8" dia. (3.175910 specimen holders For SEM 10-14 mm) pin.
All adapters have a threaded portion 5 mm in length. Adapter measurements are length x diameter.
Ordering information: Order the Adapter Pin that fits the instrument.
Product Family Options
Product Information
  • Désignation
    Ø
    Hauteur
    Épaisseur
    Availability
    Price
  • ISI First Stage Holder
    10 mm
    5 mm
    Log in for availability
    Log in for availability
    Each
    Each - 99,72 $
    99,72 $
    Each
       99,72 $
    Supplier Number: 75925
  • 3 Pin Holder, Flat Base
    25 mm
    10 mm
    Log in for availability
    Log in for availability
    Each
    Each - 238,00 $
    238,00 $
    Each
       238,00 $
    Supplier Number: 75920
  • 3 Pin Holder, With Pin B
    25 mm
    10 mm
    Log in for availability
    Log in for availability
    Each
    Each - 252,36 $
    252,36 $
    Each
       252,36 $
    Supplier Number: 75920-02
  • 5-Pin Configuration Holder With Pin A
    25 mm
    10 mm
    Discontinued
    Supplier Number: 75921-01
  • 5-Pin Configuration Holder With Pin C
    25 mm
    10 mm
    Log in for availability
    Log in for availability
    Each
    Each - 252,73 $
    252,73 $
    Each
       252,73 $
    Supplier Number: 75921-03
  • Amray Base Universal Holder
    Log in for availability
    Log in for availability
    Each
    Each - 739,10 $
    739,10 $
    Each
       739,10 $
    Supplier Number: 75910
  • Amray Base Universal Holder With Pin A
    Discontinued
    Supplier Number: 75910-01
  • Amray Base Universal Holder With Pin C
    Log in for availability
    Log in for availability
    Each
    Each - 652,54 $
    652,54 $
    Each
       652,54 $
    Supplier Number: 75910-03
  • Vertical for Thin Sample With Pin A
    25 mm
    10 mm
    Log in for availability
    Log in for availability
    Each
    Each - 478,88 $
    478,88 $
    Each
       478,88 $
    Supplier Number: 75915-01
  • Vertical for Thin Sample With Pin C
    25 mm
    10 mm
    Discontinued
    Supplier Number: 75915-03