Opaque White, Extra Fine Pen, Electron Microscopy Sciences
Product Details & Documents
Permanently mark on nearly any surface in white color and the marking is waterproof
This pen is ideal for SEM, marking sample identification on carbon tabs, aluminum stubs, conductive tape or any similar surface. The pen contains xylene.
Spécifications
- Désignation:Opaque White Pen