Spec-Cloth, Electron Microscopy Sciences | Avantor
Spec-Cloth, Electron Microscopy Sciences
Spec-Cloth, Electron Microscopy Sciences
Certificates
Spec-Cloth, Electron Microscopy Sciences
Spec-Cloth, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A long napped rayon flock with a plastic barrier.
This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Available with Pressure Sensitive Adhesive (PSA) with and without hole.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Spec-Cloth Polishing Cloth, PSA no hole
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $114.31
    $114.31
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $85.70
    $85.70
    Each
       $114.31
    Pack of 5
       $85.70
    Supplier Number: 50320-05
  • Spec-Cloth Polishing Cloth, PSA, 21/2" centered hole
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $117.83
    $117.83
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $88.18
    $88.18
    Each
       $117.83
    Pack of 5
       $88.18
    Supplier Number: 50320-15