Slide Staining Rack; Polypropylene, Electron Microscopy Sciences
Product Details & Documents
This slide staining rack is made from polypropylene and is autoclavable.
- Accommodates up to 20 slides 76×26 mm (3×1")
- Comes with a removable snap-on handle
- Measures: 70×86×21 mm (2¾×3⅜×¹³⁄₁₆") (without handle)
Spécifications
- Désignation:Slide Staining Rack