Slide Staining Dish for 20 Slides, Electron Microscopy Sciences
About this item
Ideal for immersing slides in several solutions.
The removable glass slide rack has an open bottom which allows for rapid filling and draining. It holds 20 slides-75 x 25 mm (3 x 1"), 75 x 38 mm (3 x 1 1/2"), or 75 x 51 mm (3 x 2") in size. The staining dish measures L x W x H: 107 x 87 x 70 mm. Slide Rack measures: 102 x 80 x 24 mm.
Ordering information: The complete set consists of 1x Staining dish, 1x Cover, 1x Slide Rack, and Handle.
6 Options Available Below
Product Details & Documents
Ideal for immersing slides in several solutions.
The removable glass slide rack has an open bottom which allows for rapid filling and draining. It holds 20 slides-75 x 25 mm (3 x 1"), 75 x 38 mm (3 x 1 1/2"), or 75 x 51 mm (3 x 2") in size. The staining dish measures L x W x H: 107 x 87 x 70 mm. Slide Rack measures: 102 x 80 x 24 mm.
Ordering information: The complete set consists of 1x Staining dish, 1x Cover, 1x Slide Rack, and Handle.
Product Family Options
Product Information
- DésignationAvailabilityPrice
- Supplier Number: 70312-23
- Supplier Number: 70312-25
- Supplier Number: 70312-20
- Supplier Number: 70312-24
- Supplier Number: 70312-21
- Supplier Number: 70312-22