Slide Staining Dish, Electron Microscopy Sciences | Avantor
Slide Staining Dish, Electron Microscopy Sciences
Staining Dish, 81 x 101 x 86.5 mm (3 1/8 x 4 x 3 3/8")
Catalog # 100492-958
Fournisseur #:  70321-20
Slide Staining Dish, Electron Microscopy Sciences
Catalog # 100492-958
Slide Staining Dish, Electron Microscopy Sciences
Staining Dish, 81 x 101 x 86.5 mm (3 1/8 x 4 x 3 3/8")
Price
$178.95
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The dish is made from polymethylpentene "glass clear" for use with our Polypropylene staining rack.
It comes with two lids; one lid should be used when the rack is not inserted.
Dimensions: 81 x 101 x 86.5 mm (3 1/8 x 4 x 3 3/8").
Spécifications
  • Désignation:
    Staining Dish
  • Dimensions:
    81 x 101 x 86.5 mm (3 1/8 x 4 x 3 3/8")