Slide Holder Block, Electron Microscopy Sciences | Avantor
Slide Holder Block, Electron Microscopy Sciences
90 Slide Block Holder
Catalog # 100493-394
Fournisseur #:  71532-90
Slide Holder Block, Electron Microscopy Sciences
Catalog # 100493-394
Slide Holder Block, Electron Microscopy Sciences
Slide Holder Block, Electron Microscopy Sciences
90 Slide Block Holder
Price
$362.13
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from solid high density polypropylene blocks with 13 slanted channels (on a 10° angle).
Measures W x D: 1.5 x 9 mm (1/16 x 3/8"). It is made to hold 75 x 25 mm (3 x 1") slides during cleaning, draining excess immersion oil, and even drying. They are white in color and are chemically resistant. Available in 40 or 90 capacity (vertically held). 
Spécifications
  • Désignation:
    90 Slide Block Holder