Flat Embedding Molds, Electron Microscopy Sciences | Avantor
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embedding Molds, Electron Microscopy Sciences
Certificates
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embedding Molds, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath.
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Small Flat Embedding Mold
    Discontinued
    Supplier Number: 70903
  • Flat Embed. Mold Triple Ends
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $50.52
    $50.52
    Each
       $50.52
    Supplier Number: 70875
  • Flat Embed. Mold Double Ends
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.08
    $32.08
    Each
       $32.08
    Supplier Number: 70870
  • Flat Embed Mold Single End, 4 mm (D)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.92
    $39.92
    Each
       $39.92
    Supplier Number: 70900
  • Flat Embed Mold Single End, 6 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $43.21
    $43.21
    Each
       $43.21
    Supplier Number: 70901