Eyepiece Graticules, Single Lines, Electron Microscopy Sciences
Eyepiece Graticules, Single Lines, Electron Microscopy Sciences
Certificates
Eyepiece Graticules, Single Lines, Electron Microscopy Sciences
Eyepiece Graticules, Single Lines, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Used for measurement of large objects in conjunction with a graduated mechanical stage, and for alignment as well.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • NE50 Graticule
    16 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $314.90
    $314.90
    Each
       $314.90
    Supplier Number: 67982-16
  • NE50 Graticule
    19 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $298.54
    $298.54
    Each
       $298.54
    Supplier Number: 67982-19