About this item
These test specimens are excellent for comparing magnification and assessing any distortion in the image field. It is ideally useful in the context of automated counting systems to check for unsuspected distortions.
- Made of a 5×5 mm square of single crystal silicon
- Photo-etched and the squares repeat every 10 µm
- The dividing lines are 1.9 µm wide
A broader etching line is written every 500 µm, which is useful in light microscopy. Where critical measurements must be made the sample can be mounted directly onto the calibration specimen so that an internal calibration is obtained on the micro-graph.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.