ContactUsLinkComponent
SEM, Finder Grids, Electron Microscopy Sciences
Catalog # 100495-320
Minority or Woman-Owned Business Enterprise
undefined
SEM, Finder Grids, Electron Microscopy Sciences
Catalog # 100495-320
Supplier Number:  80100-NI
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Scanning Electron Microscope SEM Finder Grid
  • Material:
    Nickel
  • Environmentally Preferable:
  • Cat. no.:
    100495-320
  • Supplier No.:
    80100-NI
Specifications
About this item
Finder grids for SEM; a valuable tool for analytical studies. The grids are available in 10 mm diameter and they can be placed directly on the stub surface and used to identify the area of interest in which the studies are performed.