Seeker/Probe, Electron Microscopy Sciences
Seeker/Probe, Electron Microscopy Sciences
Seeker/Probe, Electron Microscopy Sciences
Seeker/Probe, Electron Microscopy Sciences
Stainless steel with a hexagonal handle
Single end probe which comes with one fine point curved tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip.
Product Family Options
  • Désignation
  • Single-End Seeker/Probe
    Each
    Each - $24.61
    $24.61
    Each
       $24.61
     : 72945-01
  • Double End Seeker/Probe
    Each
    Each - $27.75
    $27.75
    Each
       $27.75
     : 72945-02