Scotch Conductive Tapes, Electron Microscopy Sciences
Scotch Conductive Tapes, Electron Microscopy Sciences
Certificates
Scotch Conductive Tapes, Electron Microscopy Sciences
Scotch Conductive Tapes, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Copper and aluminum conductive tapes with continuous long term usage up to 155 °C
  • Functional days-weeks usage up to 175 °C
  • Copper double sided available
We offer two types of foil: Aluminum foil tape (Scotch 1170) and copper foil tape (Scotch 1181) with conductive adhesive. 3” core, 1/4" (6.35 mm) width×54 ft (16.5 m) long.
Product Family Options
Informations sur le produit
  • Désignation
    Largeur
    Availability
    Price
  • Conductive Aluminum Tape
    ¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $89.55
    $89.55
    Each
       $89.55
    Supplier Number: 77800
  • Double Sided Conductive Copper Tape
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $213.74
    $213.74
    Each
       $213.74
    Supplier Number: 77802-22
  • Conductive Copper Tape
    ¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.29
    $87.29
    Each
       $87.29
    Supplier Number: 77802
  • Aluminum Tape
    1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.94
    $290.94
    Each
       $290.94
    Supplier Number: 77798
  • Aluminum Tape
    ½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $169.22
    $169.22
    Each
       $169.22
    Supplier Number: 77799
  • Copper Tape
    ½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $188.90
    $188.90
    Each
       $188.90
    Supplier Number: 77801