Scintillation Vial Rack, Electron Microscopy Sciences
À propos de cet article
Welded polypropylene rack holds 20 ml scintillation vials.
- Autoclavable
- Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
4 Options Available Below
Product Details & Documents
Welded polypropylene rack holds 20 ml scintillation vials.
- Autoclavable
- Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
Product Family Options
Informations sur le produit
- No. of PlacesW×D×HDésignationAvailabilityPrice
- Supplier Number: 72637
- Supplier Number: 72635
- Supplier Number: 72636
- Supplier Number: 72639