Stage Graticules and Micrometers for Transmitted Light, Electron Microscopy Sciences
The stage graticules are for the routine calibration of eyepiece patterns when alternating between objectives or using the graticule in different microscopes.
- Robust construction
- Ideal for student use and for instructional purposes
The scale or grid is centered on a glass disc mounted in a black anodized aluminum slide 75×24×2 mm thick.
The stage graticules are for the routine calibration of eyepiece patterns when alternating between objectives or using the graticule in different microscopes.
- Robust construction
- Ideal for student use and for instructional purposes
The scale or grid is centered on a glass disc mounted in a black anodized aluminum slide 75×24×2 mm thick.
Product Family Options
- DésignationLongueurSubdivisionsType
- Stage Graticules S110.0 mm100 Divisions (Each 0.100 mm)HorizontalEach$681.14: 68040-01
- Stage Graticules S25.0 mm100 Divisions (Each 0.050 mm)HorizontalEach$736.41: 68039-02
- Stage Graticules S42.5 mm100 Divisions (Each 0.025 mm)Horizontal-: 68039-04
- Stage Graticules S110.1 mm50 Divisions (Each 0.0025 mm)HorizontalEach$1,092.26: 68039-11
- Stage Graticules S161.0 mm100 Divisions (Each 0.010 mm)Crossed ScaleEach$1,070.93: 68039-16
- Stage Graticules S20—10 Divisions (Each 0.010 mm)Double Micrometer ScaleEach$711.55: 68043-20
- Stage Graticules S215.0 mm100 Divisions (Each 0.500 mm)Micrometer ScaleEach$705.78: 68043-21
- Stage Graticules S481.0 mm100 Divisions (Each 0.010 mm)No CoverglassEach$717.50: 68041-48
- Stage Micrometer S520.0 mm200 Divisions (Each 0.010 mm)HorizontalEach$736.41: 68039-05