SaniWash, Electron Microscopy Sciences
Antimicrobial handwash, 473 ml (16 oz.)
Catalog # 100492-990
Supplier: Electron Microscopy Sciences
: 70582-16
Jump to:
Product Details & Documents
SaniWash contains gentle cleansers and special conditioners, including Aloe Vera, to wash hands without over drying or irritating the skin.
- Kills germs on contact
- Extended antimicrobial action with PCMX
- Pleasant fragrance
- Helps to control cross contamination
Conveniently packaged in pump dispenser bottles.
Specifications
- Capacité:473 ml (16 oz.)
- Désignation:Antimicrobial handwash