Sample Vials In Mini-File®; Clear, Electron Microscopy Sciences
Product Details & Documents
These vials are made from Wheaton-33R a low extractable borosilicate glass.
- Vials and caps can be autoclaved
Delivery information: The vials are supplied with a white (styrene-butadiene) rubber lined cap.
Packaging: Comes ten vials packed in a clean plastic thermoformed Mini-File and offers convenience in transporting, displaying and storing.
Spécifications
- Avec bouchon:Oui
- Capacité:2.00 ml
- Désignation:Sample vials in Mini-File