Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences | Avantor
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Certificates
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Transport vials/tubes are ideal for sample collection and storage.
  • Conical tubes with freestanding skirts
  • Tubes are autoclavable
  • Write-on area on side
Transport Vials are made from virgin polypropylene. They are available in three sizes, and come either sterile or non sterile. Graduations provide convenient and accurate references.
Transport Tubes are made from virgin polypropylene (PP) or polystyrene (PS). They also work well in Olympus automated testing equipment. Plug caps are required.
Delivery information: 5 ml and 10 ml vials are supplied with white screw cap and 50 mL vials are supplied with blue cap that has a molded-in sealing ring for a leak-proof fit. Plug caps are available in yellow color and you can choose from hollow top (HT) or hollow bottom (HB).
Product Family Options
Transport Vials
Informations sur le produit
  • Volume
    Désignation
    Sterility
    Availability
    Price
  • 5 ml
    Sample Vials with White Caps
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $393.33
    $393.33
    Log in pour la disponibilité
    Each
    Each - $393.33
    $393.33
    Case of 500
       $393.33
    Each
       $393.33
    Supplier Number: 63035-05
  • 10 ml
    Sample Vials with White Caps
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $461.92
    $461.92
    Log in pour la disponibilité
    Each
    Each - $461.92
    $461.92
    Case of 500
       $461.92
    Each
       $461.92
    Supplier Number: 63035-10
  • 50 ml
    Sample Vials with White Caps
    Sterile
    Discontinued
    Supplier Number: -
  • 5 ml
    Sample Vials with White Caps
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $735.73
    $735.73
    Log in pour la disponibilité
    Each
    Each - $735.73
    $735.73
    Case of 1,000
       $735.73
    Each
       $735.73
    Supplier Number: 63034-05
  • 10 ml
    Sample Vials with White Caps
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $583.73
    $583.73
    Log in pour la disponibilité
    Each
    Each - $583.73
    $583.73
    Case of 1,000
       $583.73
    Each
       $583.73
    Supplier Number: 63034-10
  • 50 ml
    Sample Vials with Blue Caps
    Non sterile
    Discontinued
    Supplier Number: -
Transport Tubes
Informations sur le produit
  • Volume
    Désignation
    Sterility
    Availability
    Price
  • 12 ml
    Conical Bottom with Skirt Tube, PP, No Cap
    Discontinued
    Supplier Number: -
  • 12 ml
    Conical Bottom with Skirt Tube, PS, No Cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $243.00
    $243.00
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $185.46
    $185.46
    Each
       $243.00
    Pack of 1,000
       $185.46
    Supplier Number: 63036-PS
  • 12 ml
    Conical Bottom Tube, PP, No Cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $211.00
    $211.00
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $161.03
    $161.03
    Each
       $211.00
    Pack of 1,000
       $161.03
    Supplier Number: 63037-PP
  • 12 ml
    Flat Bottom Tube, PS, No Cap
    Discontinued
    Supplier Number: -
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Hollow Bottom Plug Cap, Yellow
    Hollow Bottom Plug Cap, Yellow
    100490-282
    Log in pour la disponibilité
    Each - $104.86
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Hollow Top Plug Cap, Yellow
    Hollow Top Plug Cap, Yellow
    100490-284
    Log in pour la disponibilité
    Each - $104.86