Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Certificates
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These lightweight transparent PVC sample bottles come with snap-on tight-fitting caps and they offer excellent clarity, durability, and good chemical resistance. Maximum temperature –20 to 80 °C (–4 to 176 °F).
Note: The following sizes are also available: 30, 40, 48, 60, 90, 120,and 180 ml.
Product Family Options
Product Information
  • Capacité
    Hauteur
    Availability
    Price
  • 30 ml (1 oz.)
    25 mm
    Discontinued
    Supplier Number: 64264-03
  • 100 ml (3.4 oz.)
    34 mm
    Discontinued
    Supplier Number: 64264-10
  • 50 ml (1.7 oz.)
    36 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 659,75 $
    659,75 $
    Log in for availability
    Pack of 50
    Pack of 50 - 499,91 $
    499,91 $
    Each
       659,75 $
    Pack of 50
       499,91 $
    Supplier Number: 64264-05