Safety Bulb, Electron Microscopy Sciences | Avantor
Safety Bulb, Electron Microscopy Sciences
Safety Bulb, Electron Microscopy Sciences
Certificates
Safety Bulb, Electron Microscopy Sciences
Safety Bulb, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Natural rubber bulb allows safe, manual control of filling or discharging toxic liquids through a pipette by finger pressure, thus avoiding mouth contact.
  • Guaranteed to be consistently flexible
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Safety Bulb
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $56.90
    $56.90
    Each
       $56.90
    Supplier Number: 70982
  • Safety Bulb
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $173.91
    $173.91
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $133.18
    $133.18
    Each
       $173.91
    Pack of 4
       $133.18
    Supplier Number: 70984