Rubber Cement, Electron Microscopy Sciences
Rubber Cement
  100491-938
Rubber Cement, Electron Microscopy Sciences
  100491-938
 :  
Rubber Cement, Electron Microscopy Sciences
Rubber Cement, Electron Microscopy Sciences
Rubber Cement
 :  
$19.80
Each
Excellent adhesive for all paper pasting and mountings of drawings and photographs. It is also used to adhere specimens to SEM Stubs. Any excess cured cement can be easily removed. 4 oz. bottle comes with a brush in the cap.
 : Certificates of analysis are available.
  • Désignation:
    Rubber Cement