WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
Certificates
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
WHEATON® 15×45 mm Shell Vials, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These vials are for use with autosamplers that require 15×45 mm vials. The vials are available in either clear or amber glass or in natural polypropylene, without 15 mm snap plug cap.
  • Conical interior for maximum sample recovery
Product Family Options
Product Information
  • Désignation
    Capacité
    Diameter ×H
    Matière
    Color
    Avec bouchon
    Availability
    Price
  • Shell Vials
    3.00 ml
    15×45 mm
    PP
    Non
    Discontinued
    Supplier Number: -
  • Shell Vials
    4.00 ml
    15×45 mm
    Glass
    Amber
    Non
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $83.10
    $83.10
    Log in for availability
    Pack of 100
    Pack of 100 - $63.77
    $63.77
    Each
       $83.10
    Pack of 100
       $63.77
    Supplier Number: 72627-4A
  • Shell Vials
    4.00 ml
    15×45 mm
    PP
    Non
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $86.03
    $86.03
    Log in for availability
    Pack of 100
    Pack of 100 - $65.74
    $65.74
    Each
       $86.03
    Pack of 100
       $65.74
    Supplier Number: 72627-4P
  • Shell Vials
    4.00 ml
    15×45 mm
    Glass
    Clear
    Non
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $87.31
    $87.31
    Log in for availability
    Pack of 100
    Pack of 100 - $66.72
    $66.72
    Each
       $87.31
    Pack of 100
       $66.72
    Supplier Number: 72627-4C
Caps
Product Information
  • Catalog #
    Désignation
    Color
    Availability
    Price
  • Snap Plug Cap for I.D.15 mm × O.D.45 mm
    Natural
    Log in for availability
    Log in for availability
    Each
    Each - $58.42
    $58.42
    Log in for availability
    Pack of 100
    Pack of 100 - $58.42
    $58.42
    Each
       $58.42
    Pack of 100
       $58.42