RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences | Avantor
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
Certificates
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
RED-Z Solidifier will solidify, decontaminate and deodorize any water base fluid at a ratio of ¾ oz to 1 L
It contains chlorine (NaDCC) an EPA registered disinfectant - almost twice the amount recommended by the CDC. Solidifier could also be used for blood and other body fluids. RED-Z available in a pointed top bottle that fits easily in porthole containers and in a flip-top shaker top bottle for large spills.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 100503-194
  • 100503-188
  • Capacity:
  • 11 oz
  • 15 oz
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • Red-Z Fluid Control Solidifier
    11 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $76.46
    $76.46
    Each
       $76.46
    Supplier Number: 24145-10
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100503-194
    Capacity
    11 oz
    Minority or Woman-Owned Business Enterprise
  • Red-Z Fluid Control Solidifier
    15 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $88.55
    $88.55
    Each
       $88.55
    Supplier Number: 24145-25
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100503-188
    Capacity
    15 oz
    Minority or Woman-Owned Business Enterprise