Quick Release Pipette Filler, Electron Microscopy Sciences | Avantor
Quick Release Pipette Filler, Electron Microscopy Sciences
Quick Release Pipette Filler, Electron Microscopy Sciences
Certificates
Quick Release Pipette Filler, Electron Microscopy Sciences
Quick Release Pipette Filler, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This pipetting device provides an easy, precise way of filling and releasing liquid.
  • Sizes are color coded
  • Fits standard or disposable pipettes and diposable serological tooled-mouthpiece pipettes as per ASTM Standard E714-80
By rotating the knurled thumb wheel the liquid is drawn up or dispensed. The soft elastic chuck has a threaded collar that secures the plastic or glass pipette to the device.
Product Family Options
Informations sur le produit
  • Volume
    Color
    Availability
    Price
  • 2 ml
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.80
    $103.80
    Each
       $103.80
    Supplier Number: 70957
  • 25 ml
    Red
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $131.69
    $131.69
    Each
       $131.69
    Supplier Number: 70959
  • Discontinued
    Supplier Number: -
  • 10 ml
    Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $105.19
    $105.19
    Each
       $105.19
    Supplier Number: 70958