Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Certificates
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Aluminum Oxide and Silicon Carbide Abrasive Cut-Off Wheels are offered for long life, and are affordably priced for various sectioning requirements.
All wheels are 228.6 mm (9") in diameter, 1.6 mm (0.062”) thick, and with 31 mm (1 1/4”) arbor.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Hard Steel Rc60+ Cut-Off Wheel
    Discontinued
    Supplier Number: -
  • Med Non-Ferrous, Ti, Cut-Off Wheel
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $484.51
    $484.51
    Log in for availability
    Pack of 5
    Pack of 5 - $406.09
    $406.09
    Each
       $484.51
    Pack of 5
       $406.09
    Supplier Number: 50209-25
  • Soft Steel Rc30-45 Cut-Off Wheel
    Discontinued
    Supplier Number: -
  • Soft Non-Ferrous, Ti, Cut-Off Wheel
    Discontinued
    Supplier Number: -
  • Hard Non-Ferrous, Ti, Cut-Off Wheel
    Discontinued
    Supplier Number: -