Precision Pin Vises, Electron Microscopy Sciences
Precision Pin Vises, Electron Microscopy Sciences
Precision Pin Vises, Electron Microscopy Sciences
Certificates
Precision Pin Vises, Electron Microscopy Sciences
Precision Pin Vises, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These tool steel collets grip firmly when screw chucks are tightened.
  • The chromium-plated handle is hollow which allows for the holding of long wires
The body is stepped, with the small portion knurled allowing for rotation between the fingers. The larger portion is smooth and it easily can be held firmly in lathes and drill presses.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Precision pin vise, 3"
    Log in for availability
    Log in for availability
    Each
    Each - $45.85
    $45.85
    Each
       $45.85
    Supplier Number: 62074-01
  • Precision pin vise, 31/4"
    Discontinued
    Supplier Number: 62074-02
  • Precision pin vise, 41/4"
    Discontinued
    Supplier Number: 62074-04
  • Precision pin vise, 4/Set
    Discontinued
    Supplier Number: 62074-10