Portable Vacu-Vises, Electron Microscopy Sciences | Avantor
Portable Vacu-Vises, Electron Microscopy Sciences
Multi-angle vacu-vise
Catalog # 100496-134
Fournisseur #:  62068
Portable Vacu-Vises, Electron Microscopy Sciences
Catalog # 100496-134
Portable Vacu-Vises, Electron Microscopy Sciences
Portable Vacu-Vises, Electron Microscopy Sciences
Multi-angle vacu-vise
Price
$191.44
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
With a rubber suction base; you just flick off the Vacu-lever and the Vacu-vise firmly fastens to any clean, smooth surface.
When you lift the Vacu-lever up, the Vacu-vise is released. The Multi-Angle Swivel Vacu-vise can be tilted on any desired angle. Swivels 360°. 3" metal jaws and it is supplied with a slide-on rubber to hold delicate objects; opens up to 2 5/8"; 61/4" triangle base.
Spécifications
  • Désignation:
    Multi-angle vacu-vise