Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Holder, Electron Microscopy Sciences
Certificates
Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Holder, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Pop-Up holder holds 20 or 30 of 1 x 3" microscope slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed. Measurements: 8W x 131/2L x 11/32H".
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Pop-Up Slide Folder
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $36.74
    $36.74
    Each
       $36.74
    Supplier Number: 71520
  • Pop-Up Slide Folder
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $323.02
    $323.02
    Each
       $323.02
    Supplier Number: 71521
  • 30 Slide Pop-Up Slide Folder
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $40.28
    $40.28
    Each
       $40.28
    Supplier Number: 71522
  • 30 Slide Pop-Up Slide Folder
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $364.65
    $364.65
    Log in pour la disponibilité
    Each
    Each - $364.65
    $364.65
    Case of 10
       $364.65
    Each
       $364.65
    Supplier Number: 71522-10