Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
Product Family Options
- CapacitéHauteurØ int.
- 5 Dram (18.5 ml)40 mm22 mmEach$37.55Pack of 10$28.20: 64248-15
- 30 Dram (111 ml)76 mm45 mmEach$57.94Pack of 10$39.99: 64248-45
- 13 Dram (48 ml)56 mm30 mm-: 64248-30
- 7 Dram (25.9 ml)51 mm26 mm-: 64248-20
- 20 Dram (74 ml)64 mm38 mm-: 64248-40
- 3 Dram (11 ml)48 mm19 mmEach$37.55Pack of 10$28.20: 64248-10
- 40 Dram (148 ml)102 mm45 mmEach$55.23Pack of 10$34.32: 64248-50
- 15 Dram (55.5 ml)64 mm32 mmEach$42.82Pack of 10$34.27: 64248-35