Polyethylene Vials; Poly-Vials, Electron Microscopy Sciences
À propos de cet article
These flat bottom cylindrical vials have a a friction-fit, flip-top closure which can not be lost. Ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
- Unbreakable
- Withstand temps up to 90 °C
- Wall thickness 1 mm
11 Options Available Below
Product Details & Documents
These flat bottom cylindrical vials have a a friction-fit, flip-top closure which can not be lost. Ideal for use as a specimen container for the storage of small samples, or as embedding capsules.
- Unbreakable
- Withstand temps up to 90 °C
- Wall thickness 1 mm
Product Family Options
Informations sur le produit
- DésignationCapacitéDiameter ×HAvec bouchonAvailabilityPrice
- Poly-vial1.00 ml9×32 mmOuiEach$25.38Pack of 10$19.24Supplier Number: 72621-10
- Poly-vial1.00 ml9×32 mmOuiCase of 100$212.45Each$212.45Supplier Number: 72621-12
- Poly-vial2.50 ml12×40 mmOuiEach$28.59Pack of 10$22.04Supplier Number: 72621-20
- Poly-vial2.50 ml12×40 mmOuiCase of 100$262.85Each$262.85Supplier Number: 72621-22
- Poly-vial5.00 ml15×50 mmOuiEach$26.44Pack of 10$20.24Supplier Number: 72621-30
- Poly-vial5.00 ml15×50 mmOuiCase of 100$264.47Each$264.47Supplier Number: 72621-32
- Poly-vial8.00 ml17×56 mmOuiCase of 100$301.41Each$301.41Supplier Number: 72621-38
- Poly-vial10.00 ml22×32 mmOuiEach$38.60Pack of 10$29.97Supplier Number: 72621-40
- Poly-vial10.00 ml22×32 mmOuiCase of 100$354.18Each$354.18Supplier Number: 72621-42
- Poly-vial25.00 ml24×72 mmOuiEach$75.96Pack of 10$53.94Supplier Number: 72621-60
- Poly-vial50.00 ml30×74 mmOuiCase of 100$805.61Each$805.61Supplier Number: 72621-92