Plastic Tissue Capsules, Electron Microscopy Sciences | Avantor
Plastic Tissue Capsules, Electron Microscopy Sciences
Plastic Tissue Capsule, 35 I.D. x 10 mm Depth, 35×10 mm
Catalog # 100490-156
Fournisseur #:  62322-12
Plastic Tissue Capsules, Electron Microscopy Sciences
Catalog # 100490-156
Plastic Tissue Capsules, Electron Microscopy Sciences
Plastic Tissue Capsules, Electron Microscopy Sciences
Plastic Tissue Capsule, 35 I.D. x 10 mm Depth, 35×10 mm
Price
$236.17
Log in pour la disponibilité
Each Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
They measure 35 mm (I.D.) x 35 mm (I.D.) x 10 mm (deep) (1 3/8" x 5/16").
  • Molded from polyethylene; resistant to all processing chemicals, including Zenker's fixative.
  • A three-compartment divider which is removable for specimens up to 35mm.
  • A weighted end floats the capsule in a upright position to conserve space.
  • Perforated on the top and bottom for better circulation of the fluid.
  • Snap-on hinged cover.
Spécifications
  • Désignation:
    Plastic Tissue Capsule
  • Dimensions:
    35 I.D. x 10 mm Depth
  • Diameter ×H:
    35×10 mm