Graduated Transfer Pipettes, Electron Microscopy Sciences
À propos de cet article
These disposable graduated transfer pipettes are made up of polyethylene.
- Low-affinity surface
- Attached bulb avoids cross-contamination
- One step biohazard disposable
These pipettes are ideal for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, etc.
7 Options Available Below
Product Details & Documents
These disposable graduated transfer pipettes are made up of polyethylene.
- Low-affinity surface
- Attached bulb avoids cross-contamination
- One step biohazard disposable
These pipettes are ideal for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, etc.
Product Family Options
Sterile
Informations sur le produit
- DésignationCapacitéØ×LGraduationBulb DrawPackedAvailabilityPrice
- Graduated Transfer Pipettes1.5 ml3.0×11.6 cm0.3 mL0.8 mlIndividually PackedEach$200.69Pack of 500$144.06Supplier Number: 70962-11S
- Graduated Transfer Pipettes5.8 ml4.6×15.5 cm1 mL3.4 mlIndividually PackedEach$217.78Pack of 500$161.62Supplier Number: 70962-8S
Non-sterile
Informations sur le produit
- DésignationCapacitéØ×LGraduationBulb DrawPackedAvailabilityPrice
- Graduated Transfer Pipettes1.5 ml3.0×11.6 cm0.3 mL0.8 mlBulkEach$95.85Pack of 500$70.78Supplier Number: 70962-11
- Graduated Transfer Pipettes3.9 ml4.7×14 cm1 mL2.2 mlBulkEach$81.11Pack of 500$62.38Supplier Number: 70962-7
- Graduated Transfer Pipettes4.8 ml7.1×15.2 cm2 mL1.8 mlBulkEach$80.57Pack of 500$60.69Supplier Number: 70962-10
- Graduated Transfer Pipettes5.8 ml4.6×15.5 cm1 mL3.4 mlBulkEach$89.40Pack of 500$67.17Supplier Number: 70962-8
- Graduated Transfer Pipettes7.5 ml7.9×15.5 cm3 mL3.2 mlBulkEach$95.49Pack of 500$72.60Supplier Number: 70962-9