G57 Pharmaceutical PSA Patterns, Electron Microscopy Sciences
This graticule was designed for the pharmaceutical industry.
- Scale is incorporated
- Glass sandwich surface
However, it is also useful where particle size considerations are restricted to 10 and 25 mm. Dots and circles give quick references for these two sizes.
The microscope must be calibrated when ordering this graticule, such that the circle must equate to one mm on the microscope stage.
This graticule was designed for the pharmaceutical industry.
- Scale is incorporated
- Glass sandwich surface
However, it is also useful where particle size considerations are restricted to 10 and 25 mm. Dots and circles give quick references for these two sizes.
The microscope must be calibrated when ordering this graticule, such that the circle must equate to one mm on the microscope stage.
Product Family Options
- DésignationTaille
- : 68091-20.4
- : 68091-23
- : 68091-24.5MarketsourceMarketsource
- : 68091-25
- : 68091-27